Anti-OVA Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4633S 200-4633S
RL2004633SEA 183.25 USD
RL2004633S CA11029-044
Anti-OVA Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: OVA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR