Anti-DEX Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-406-089S 200-406-089S
RL200406089SEA 147.5 USD
RL200406089S CA11029-204
Anti-DEX Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: DEX
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Penicillium
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR