Anti-IDO1 Mouse Monoclonal Antibody [clone: 10.1]

Supplier: Rockland Immunochemicals
200-301-E57S
RL200301E57SEA 147.5 USD
RL200301E57S
Anti-IDO1 Mouse Monoclonal Antibody [clone: 10.1]
Antibodies
Type: Primary
Antigen: IDO1
Clonality: Monoclonal
Clone: 10.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR