L15 Paraffin Filtering Automats, Medite
02-1800-00 46-1801-00 02-1810-00
10014-750EA 8333.18 USD
10014-750 10768-984 10768-986
L15 Paraffin Filtering Automats, Medite
The L15 paraffin filtering automats are designed to melt, filtrate, and recycle fresh and used paraffin.
  • Multiple advantages for histology laboratories
  • Melts down old blocks
  • Obtained, filtered paraffin can be reused
This unique instrument presents multiple advantages for the routine histology laboratory. It serves to avoid the complicated and time-consuming melting of paraffin using Bunsen burners, incubators, or pots, and helps to save space in the laboratory.
As the special filter cartridge retains all residue, even old blocks can be melted down. Adding the same quantity of fresh paraffin, the filtered paraffin obtained can be used again without any significant loss of quality. The large work space below the safety spigot enables easy paraffin dispensing. The unit is made of stainless steel and aluminum, therefore its surfaces are resistant to most solvents.
The operation volume of the paraffin tank is approximately 15 liters so there is always a sufficient amount of melted paraffin ready for use. Consequent application of the paraffin recycling process enables paraffin savings of up to 50%.
Ordering information: A 115V unit is also available; contact your VWR representative for more information.