TFB 55 Tissue Flotation Bath with Extra Wide Rim, Medite
01-5510-00 01-5510-00
10768-880EA 1390.18 USD
10768-880 CA10767-666
TFB 55 Tissue Flotation Bath with Extra Wide Rim, Medite
Water bath for flattening out and drying tissue sections in histology, pathology, chemical, clinical and bacteriology laboratories
The tissue flotation bath provides a circular water bath with exact mechanical temperature regulation. Its bath well and extra wide rim are made of high quality aluminum with black coating. The safe heating system with ceramic ballast resistances avoids overheating and damage even in case of an empty bath.