TES 99 Tissue Embedding System, Medite
02-9970-00 02-9951-00 02-9970-00 02-9935-00 02-9952-00 02-9952-00 02-9951-00 02-9935-00
10768-804EA 4991.9 USD
10768-804 10768-706 10768-806 CA10767-682 CA10767-584 CA10767-680 CA10767-586 10768-802
TES 99 Tissue Embedding System, Medite
Thermal unit with deep paraffin tray (up to 75 °C / 167 °F) that accepts baskets from customary tissue processors, preheating container temperature up to 70 °C / 158 °F.
  • Dispensing unit with paraffin tank of 0.8 gal. (3.5 liters)
  • Cold spot and heat protective hand rest
  • Made of stainless steel
  • Large unit cools down to -15 °C / 5 °F
Available with large cooling Unit with storage space for 110 paraffin blocks or with a small cooling unit of compact size for space-limited laboratories for up to 80 blocks.