Target™ Snap-It™ Vial Storerooms, Thermo Scientific
C4075-43
66030-620EA 945.56 USD
66030-620
Target™ Snap-It™ Vial Storerooms, Thermo Scientific
Thermo Scientific National Target Snap-It Storerooms provide convenient storage for vials and caps.
Mini Storeroom has 6 drawers, each drawer will hold one 100/pack of vials or 500/pack caps. Full-size Storeroom has 9 deep drawers with a capacity of 2,000 vials and caps. Storeroom can be restocked as product is consumed.