Target™ Crimp-Top Vial Storerooms, Thermo Scientific
Supplier: Thermo Scientific
C4075-21 C4075-1 C4011-688
66030-222EA 1480.15 USD
66030-222 66064-942 66030-202
Target™ Crimp-Top Vial Storerooms, Thermo Scientific
Thermo Scientific National Storerooms with Target Crimp-Top Vials feature your choice of mini (six-drawer) or full size (nine-drawer) storage units complete with vials and caps.
Mini Storeroom has 6 drawers; each drawer contains 100 packs of vials or septa. Full size storeroom has 9 deep drawers with a capacity of 2,000 vials and seals. Stackable cabinet constructed of heavy-gauge steel with metal shelves and injection-molded drawers. Storeroom can be restocked as product is consumed. Prestocked with 3000 of each: clear vials and preassembled clear crimp seals.