Safety Tape, Black and White Striped, Micronova
SST18-2BLWH
89236-630CS 305.76 USD
89236-630
Safety Tape, Black and White Striped, Micronova
Black and white striped pressure sensitive tape for delineating and color coding specific equipment and areas.
Laminated vinyl tape is scuff resistant and holds up well to solvents, humidity and oil. Wound on a plastic core and cleanroom processed.