Isomaltotriose 97%, Powder

Supplier: Thermo Scientific Chemicals

J64186-ME J64186-MC J64186-ME J64186-MC
AAJ64186-MEEA 571.19 USD
AAJ64186-ME AAJ64186-MC CAAAJ64186-ME CAAAJ64186-MC
Isomaltotriose 97%, Powder
Isomaltotriose
Formula: C₁₈H₃₂O₁₆
MW: 504.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070623
CAS Number: 3371-50-4
EINECS: 222-149-2

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR