2-Amino-4'-methoxyacetophenone hydrochloride 95%

Warning

H66966-06 H66966-03
AAH66966-03EA 60.7 USD
AAH66966-03 AAH66966-06
2-Amino-4'-methoxyacetophenone hydrochloride 95%
2-Amino-4'-methoxyacetophenone hydrochloride
Formula: C₉H₁₂ClNO₂
Melting Pt: 190…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00193078
CAS Number: 3883-94-1

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR