N-Acetyl-D-methionine 98%

Synonyms: Ac-D-Met-OH

H66808-06 H66808-22 H66808-14
AAH66808-22EA 401.82 USD
AAH66808-22 AAH66808-14 AAH66808-06
N-Acetyl-D-methionine 98%
N-Acetyl-D-methionine
Formula: C₇H₁₃NO₃S
MW: 191.2 g/mol
Melting Pt: 102…104 °C
Storage Temperature: Freezer
MDL Number: MFCD00069962
EINECS: 216-144-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR