3-(Hydroxymethyl)-1-adamantanol 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Hydroxy-3-(hydroxymethyl)adamantane

H66721-03 H66721-MD H66721-06
AAH66721-03EA 91.29 USD
AAH66721-03 AAH66721-06 AAH66721-MD
3-(Hydroxymethyl)-1-adamantanol 96%
3-(Hydroxymethyl)-1-adamantanol
Formula: C₁₁H₁₈O₂
MW: 182.26 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD03105096
CAS Number: 38584-37-1

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR