Boc-Lys(fmoc)-OMe 95%

Synonyms: (S)-Methyl 6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoateNα-Boc-Nepsilon-Fmoc-L-lysine methyl ester

H66712-03 H66712-MD
AAH66712-03EA 145.41 USD
AAH66712-03 AAH66712-MD
Boc-Lys(fmoc)-OMe 95%
Boc-Lys(fmoc)-OMe
Formula: C₂₇H₃₄N₂O₆
MW: 482.577 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00236856
CAS Number: 133628-28-1

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR