3-Bromo-5-fluoroanthranilic acid 95%

Warning

Synonyms: 2-Amino-3-bromo-5-fluorobenzoic acid

H66628-MD H66628-03
AAH66628-MDEA 107.44 USD
AAH66628-MD AAH66628-03
3-Bromo-5-fluoroanthranilic acid 95%
3-Bromo-5-fluoroanthranilic acid
Formula: C₇H₅BrFNO₂
MW: 234.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00662509
CAS Number: 259269-84-6

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR