4'-Benzyloxy-2-bromopropiophenone 97%

Supplier: Thermo Scientific Chemicals

Warning

H66578-22 H66578-14
AAH66578-22EA 390.72 USD
AAH66578-22 AAH66578-14
4'-Benzyloxy-2-bromopropiophenone 97%
4'-Benzyloxy-2-bromopropiophenone
Formula: C₁₆H₁₅BrO₂
Melting Pt: 71…76 °C
Storage Temperature: Freezer
MDL Number: MFCD00017862
CAS Number: 35081-45-9
EINECS: 252-351-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR