(S)-4-Hydroxypyrrolidine-2-one 97+%

Warning

Synonyms: (S)-(-)-4-Hydroxy-2-pyrrolidinone(S)-4-Hydroxy-2-pyrrolidone(4S)-(-)-beta-Hydroxy-gamma-butyrolactam

H66455-MD H66455-03
AAH66455-03EA 109.99 USD
AAH66455-03 AAH66455-MD
(S)-4-Hydroxypyrrolidine-2-one 97+%
(S)-4-Hydroxypyrrolidine-2-one
Formula: C₄H₇NO₂
MW: 101.105 g/mol
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00273370
CAS Number: 68108-18-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR