Dde-Lys(Fmoc)-OH 98%

Supplier: Thermo Scientific Chemicals

Synonyms: DDE-Lys(Fmoc)-OHNα-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine

H66446-MD H66446-03
AAH66446-03EA 265.99 USD
AAH66446-03 AAH66446-MD
Dde-Lys(Fmoc)-OH 98%
Dde-Lys(Fmoc)-OH
Formula: C₃₁H₃₆N₂O₆
MW: 532.637 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00467662
CAS Number: 156648-40-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR