(S)-(-)-1-Fmoc-4-oxopiperidine-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fmoc-L-Homopro(4-oxo)-OH(S)-(-)-1-Fmoc-4-oxohomoproline

H66428-03 H66428-MD
AAH66428-03EA 188.99 USD
AAH66428-03 AAH66428-MD
(S)-(-)-1-Fmoc-4-oxopiperidine-2-carboxylic acid 97%
(S)-(-)-1-Fmoc-4-oxopiperidine-2-carboxylic acid
Formula: C₂₁H₁₉NO₅
MW: 365.38 g/mol
Melting Pt: 228…223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03094868
CAS Number: 1221793-43-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR