5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate 95%

Synonyms: N-Boc-L-glutamic Acid 5-Cyclohexyl EsterN-(tert-Butoxycarbonyl)-L-glutamic Acid 5-Cyclohexyl Ester5-Cyclohexyl N-Boc-L-glutamate

H66409-06 H66409-22 H66409-14
AAH66409-22EA 342.34 USD
AAH66409-22 AAH66409-14 AAH66409-06
5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate 95%
5-Cyclohexyl-N-(tert-butoxycarbonyl)-L-glutamate
Formula: C₁₆H₂₇NO₆
MW: 329.39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065570
CAS Number: 73821-97-3
EINECS: 616-021-9

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR