2-Amino-5-pyrimidineboronic acid 95%

Warning

Synonyms: 2-Aminopyrimidine-5-boronic acid(2-Aminopyrimidin-5-yl)boronic acid

H66403-06 H66403-03 H66403-MD
AAH66403-06EA 260.59 USD
AAH66403-06 AAH66403-MD AAH66403-03
2-Amino-5-pyrimidineboronic acid 95%
2-Amino-5-pyrimidineboronic acid
Formula: C₄H₆BN₃O₂
MW: 138.92 g/mol
Melting Pt: 206…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07375147
CAS Number: 936250-22-5

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR