2,4-Dimethoxy-4'-hydroxybenzophenone 98%

Warning

Synonyms: (2,4-Dimethoxyphenyl)(4-hydroxyphenyl)methanone

H66217-14 H66217-22 H66217-06
AAH66217-14EA 148.14 USD
AAH66217-14 AAH66217-06 AAH66217-22
2,4-Dimethoxy-4'-hydroxybenzophenone 98%
2,4-Dimethoxy-4'-hydroxybenzophenone
Formula: C₁₅H₁₄O₄
MW: 258.27 g/mol
Melting Pt: 138…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00137850
CAS Number: 41351-30-8

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR