tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-BOC-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester2-Methyl-2-propanyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-1(2H)-pyridinecarboxylate1(2H)-Pyridinecarboxylic acid, 3,6-dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-, 1,1-dimethylethyl ester

H66193-14 H66193-06 H66193-03
AAH66193-14EA 1055.99 USD
AAH66193-14 AAH66193-03 AAH66193-06
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate 95%
tert-Butyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate
Formula: C₁₆H₂₈BNO₄
MW: 309.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840345
CAS Number: 286961-14-6

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR