(+)-3-Bromocamphor 98%

Warning

Synonyms: (+)-Camphor Bromide

H66011-36 H66011-22
AAH66011-36EA 305.25 USD
AAH66011-36 AAH66011-22
(+)-3-Bromocamphor 98%
(+)-3-Bromocamphor
Formula: C₁₀H₁₅BrO
MW: 231.13 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00003744
CAS Number: 10293-06-8
EINECS: 233-652-1
UN: 1759
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR