(R)-4-Hydroxypyrrolidin-2-one 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-(+)-4-Hydroxy-2-pyrrolidinone
H66006-MD
H66006-03
AAH66006-MDEA
45.39
USD
AAH66006-MD
AAH66006-03
(R)-4-Hydroxypyrrolidin-2-one 97+%
(R)-4-Hydroxypyrrolidin-2-one
Formula:
C₄H₇NO₂ MW: 101.11 g/mol Melting Pt: 155…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00273364 CAS Number: 22677-21-0 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...