D-Proline benzyl ester hydrochloride 97%

Synonyms: H-D-Pro-OBzl.HCl

H65804-06 H65804-03
AAH65804-06EA 281.31 USD
AAH65804-06 AAH65804-03
D-Proline benzyl ester hydrochloride 97%
D-Proline benzyl ester hydrochloride
Formula: C₁₂H₁₅NO₂
MW: 241.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153458
CAS Number: 53843-90-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR