(S)-2-Amino-2-phenylacetamide hydrochloride 97%

Warning

Synonyms: H-Phg-NH₂·HCl

H65751-06 H65751-03
AAH65751-03EA 53.63 USD
AAH65751-03 AAH65751-06
(S)-2-Amino-2-phenylacetamide hydrochloride 97%
(S)-2-Amino-2-phenylacetamide hydrochloride
Formula: C₈H₁₁ClN₂O
MW: 186.639 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270568
CAS Number: 60079-51-8

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR