3-(Bromomethyl)thiophene 97%

Warning

H61497-06
AAH61497-06EA 382.32 USD
AAH61497-06
3-(Bromomethyl)thiophene 97%
3-(Bromomethyl)thiophene
Formula: C₅H₅BrS
Boiling Pt: 76 °C/1 mm
Density: 1.616
Storage Temperature: Ambient
MDL Number: MFCD03840497
CAS Number: 34846-44-1
EINECS: 252-247-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR