Lutetium tribromide, ultra dry ≥99.99% (metals basis), powder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lutetium (III) bromide

ampouled under argon

Ultra Dry
047190-03 047190-06
AA47190-03EA 96.79 USD
AA47190-03 AA47190-06
Lutetium tribromide, ultra dry ≥99.99% (metals basis), powder
Lutetium tribromide
Formula: Br₃Lu
MW: 414.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049965
CAS Number: 14456-53-2
EINECS: 238-446-5

Order Now

Specification Test Results

Assay Minimum 99.99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR