Cobalt bis(trifluoromethylsulfonyl)imide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bis(trifluoromethane)sulfonimide cobalt salt
047053-06
047053-03
AA47053-03EA
125.99
USD
AA47053-03
AA47053-06
Cobalt bis(trifluoromethylsulfonyl)imide
Cobalt bis(trifluoromethylsulfonyl)imide
Solid.
Formula:
C₄CoF₁₂N₂O₈S₄ MW: 619.23 g/mol Melting Pt: >200 °C Storage Temperature: Ambient |
MDL Number:
MFCD23380169 CAS Number: 207861-61-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...