Minocycline hydrochloride 50 mg/ml in distilled water, sterile filtered

J67469-XF
AAJ67469-XFEA 140.89 USD
AAJ67469-XF
Minocycline hydrochloride 50 mg/ml in distilled water, sterile filtered
Minocycline hydrochloride

Minocycline hydrochloride, 50 mg/ml in distilled water, sterile-filtered

Formula: C₂₃H₂₇N₃O₇
Storage Temperature: Refrigerator
MDL Number: MFCD00083669
CAS Number: 13614-98-7
EINECS: 237-099-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR