Monensin monosodium salt 50 mg/ml in methanol, sterile filtered

Danger

Synonyms: Monensin, Sodium SaltMonensin sodium salt

J67431-XF
AAJ67431-XFEA 66.72 USD
AAJ67431-XF
Monensin monosodium salt 50 mg/ml in methanol, sterile filtered
Monensin monosodium salt

Monensin sodium salt, 50 mg/ml in methanol, sterile-filtered

Formula: C₃₆H₆₁NaO₁₁
MW: 692.85 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077826
CAS Number: 22373-78-0
EINECS: 244-941-7
UN: 1230
ADR: "3",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR