Cellotetraose 95%, Powder
Supplier: Thermo Scientific Chemicals
Synonyms:
D-(+)-Cellotetraose
, Glc beta(1-4)Glc beta(1-4)Glc beta(1-4)Glc
Derived from cellulose degradation. Used by cellulosic bacteria as an energy source. Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidases.
Applications: Derived from cellulose degradation. Used by cellulosic bacteria as an energy source. Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and -glucosidases.
Formula:
C₂₄H₄₂O₂₁ MW: 666.58 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00079079 CAS Number: 38819-01-1 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...