3-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 95%

Warning

Synonyms: 3-(Dimethylcarbamoyl)-2-Fluorobenzeneboronic Acid

H56087-MD H56087-MD H56087-03 H56087-03
AAH56087-MDEA 82.81 USD
AAH56087-MD CAAAH56087-MD AAH56087-03 CAAAH56087-03
3-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 95%
3-(Dimethylcarbamoyl)-2-fluorophenylboronic acid
Formula: C₉H₁₁BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD16295256
CAS Number: 1451391-88-0

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR