Ceramic 2 Bore Tubes, AL23, Oval Cross Section Insulators, Thermo Scientific
Supplier: Thermo Scientific
32775-QAT 32775-DM 32775-DW 32775-DR 32775-DW 32775-DR 32775-DM 32775-QAT
AA32775-DMEA 23.85 USD
CAAA32775-DR AA32775-DM CAAA32775-QAT AA32775-DR AA32775-QAT AA32775-DW CAAA32775-DW CAAA32775-DM
Ceramic 2 Bore Tubes, AL23, Oval Cross Section Insulators, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.