Anisomycin ≥97% (by TLC)
Supplier: Enzo Life Sciences
BML-ST102-0050
89164-784EA
255.63
USD
89164-784
Anisomycin ≥97% (by TLC)
Anisomycin
Activator of MAPK, Jnk and p38
Isolated from Streptomyces griseolus.
Antibiotic. Activates JNK/SAPKs, p38 and MAP kinases but not ERKs leading to the phosphorylation of S6, histone H3 and HMG-14 and the induction of the immediate-early genes c-fos and c-jun. Induces apoptosis in the human monoblastoid cell line. Used at 300 ng/ml. Inhibits peptidyl transferase at higher concentrations. Used in the eradication of bean mildew. Inhibits other pathogenic fungi in plants.
Formula:
C₁₄H₁₉NO₄ |
MDL Number:
MFCD00077650 CAS Number: 22862-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...