Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SHURMARK® Routine Color Microscope Slides, Triangle Biomedical

Supplier: General Data Healthcare
SHUR/Mark®
SM-CS-45G SM-CS-BY SM-CS-45B SM-CCS-BY SM-CCS-BW SM-CCS-W SM-CCS-45P SM-CCS-Y SM-CCS-P SM-CS-G SM-CCS-45Y SM-CS-B SM-CCS-45W SM-CCS-G SM-CS-P SM-CCS-45B SM-CCS-BB SM-CCS-L SM-CS-L SM-CS-BB SM-CS-W SM-CCS-45L SM-CS-Y SM-CS-BG SM-CCS-B SM-CCS-45G SM-CS-45Y SM-CS-45W SM-CCS-BP SM-CS-BL SM-CCS-BL SM-CS-BP SM-CS-45P SM-CCS-BG SM-CS-BW SM-CS-45L
SHURMark Routine Color Microscope Slides are ground and polished and available in six popular colors

They can be used with pens and pencils.



 

ORDER

Expand All / Collapse All
Description L x W Thickness Color Supplier No. VWR Catalog Number Unit Price Quantity
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm White SM-CCS-45W 89413-684 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Blue SM-CCS-45B 89413-686 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Pink SM-CCS-45P 89413-690 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Yellow SM-CCS-45Y 89413-692 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Lilac SM-CCS-45L 89413-694 Case of 1,440 Retrieving
Charged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Green SM-CCS-45G 89413-688 Case of 1,440 Retrieving
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Blue SM-CCS-B 89413-662 Case of 1,440 Retrieving
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Green SM-CCS-G 89413-664 Case of 1,440 Retrieving
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Pink SM-CCS-P 89413-666 Case of 1,440 Retrieving
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Yellow SM-CCS-Y 89413-668 Case of 1,440 Retrieving
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Lilac SM-CCS-L 89413-670 Case of 1,440 Retrieving
Charged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm White SM-CCS-W 89413-660 Case of 1,440 Retrieving
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Lilac SM-CCS-BL 89413-718 Case of 1,440 Retrieving
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm White SM-CCS-BW 89413-708 Case of 1,440 Retrieving
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Green SM-CCS-BG 89413-712 Case of 1,440 Retrieving
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Yellow SM-CCS-BY 89413-716 Case of 1,440 Retrieving
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Pink SM-CCS-BP 89413-714 Case of 1,440 Retrieving
Charged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Blue SM-CCS-BB 89413-710 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm White SM-CS-45W 89413-672 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Blue SM-CS-45B 89413-674 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Yellow SM-CS-45Y 89413-680 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Pink SM-CS-45P 89413-678 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Green SM-CS-45G 89413-676 Case of 1,440 Retrieving
UnCharged with 45° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Lilac SM-CS-45L 89413-682 Case of 1,440 Retrieving
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm White SM-CS-W 89413-648 Case of 1,440 Retrieving
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Lilac SM-CS-L 89413-658 Case of 1,440 Retrieving
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Blue SM-CS-B 89413-650 Case of 1,440 Retrieving
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Pink SM-CS-P 89413-654 Case of 1,440 Retrieving
UnCharged with 90° Clipped Corner 75 x 25 mm (3 x 1") 1 mm Green SM-CS-G 89413-652 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Blue SM-CS-BB 89413-698 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Yellow SM-CS-BY 89413-704 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Lilac SM-CS-BL 89413-706 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Pink SM-CS-BP 89413-702 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm Green SM-CS-BG 89413-700 Case of 1,440 Retrieving
UnCharged with Beveled Corner 75 x 25 mm (3 x 1") 1 mm White SM-CS-BW 89413-696 Case of 1,440 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty