Would you like to log in again?
Anti-HSP60 IgG/A/M (human), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
EKS-650
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.
For the quantitative determination of anti-HSP60 IgG/A/M autoantibodies in human serum.
This assay allows for reproducible, accurate and precise determination of IgG , IgA and IgM antibodies (total) to human Hsp60 in serum. The ELISA kit uses recombinant human Hsp60 bound to the wells of the plate to bind anti-human Hsp60 antibodies present in human serum. The captured anti-human Hsp60 antibodies are detected with a HRP conjugated goat polyclonal antibody specific for human IgG, IgA and IgM molecules. The assay is developed with tetramethylbenzidine substrate pro ducing a blue color in proportion to the amount of captured anti -human Hsp60 antibodies. The color development is stopped with acid stop solution which converts the endpoint color to yellow. The intensity of the color is measured in a microplate reader at 450 nm.
Ordering information: Contains Microtiter plate, Standard, Sample diluent 1, Wash buffer concentrate, HRP Conjugate, TMB Substrate, Stop solution 2.
ORDER
Expand All / Collapse All
No. of Wells Supplier No. VWR Catalog Number Unit Price Quantity
96 Wells ADI-EKS-650 89141-856 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Sensitivity 2.88 ng/ml (range 7.81 - 250 ng/ml)
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty