Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Replicator 5th Generation Desktop 3D Printer

Supplier: D&H
MP05825 MP06077 MP06165 MP06681 MP06585 MP06584
Replicator 5th Generation Desktop 3D Printer brings the power of rapid prototyping and engineering into the classroom.

  • Easy-To-Load Filament
  • Simple, Interactive User Panel
  • Paper-Thin Layers on Models
  • Models Have Smooth Surfaces That Don’t Need Sanding, Finishing, or Postproduction
  • Realistic Prototypes and Models for Demonstrations and Presentations

The printer is powered by the new MakerBot Replicator 3D Printing Platform. The MarkerBot’s features include the Smart Extruder, motion controllers, and gantry design.

App and cloud enabled Wi-Fi, USB, and Ethernet connectivity ensures a seamless production workflow. The printer features on-board camera and diagnostics and assisted build plate leveling.

Ordering information: Makerbot filament is sold separately.



 

ORDER

Expand All / Collapse All
Description Input Voltage Supplier No. VWR Catalog Number Unit Price Quantity
Build Plate Tape 120-240V AC MP06077 470160-912 Each Retrieving
CSA Certified
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Weight 41 Kg (90 lbs.)
L×W×H 49.3 x 56.7 x 86.1 cm
Build Volume 25.2L x 19.9W x 15H cm (9.9 x 7.8 x 5.9"); 7522 cu. cm (456 cu. in.)

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping