Nor-Lake® Scientific Humidity Stability Chamber-Refrigerated Incubator, Digital 132 x 64 LED display, Programmable logic (PLC) microprocessor temperature controller, Audible and visual Hi/Lo temperature alarm, Adjustable temperature and humidity control range: +4°C to +70°C, Humidity transmitter (0-10 VDC), Temperature transmitter (0-10 VDC) (selectable to air temperature), Programmable ramp and soak, Remote alarm contacts, Continuous product temperature display, 3 sensor system, 1 product, 1 air, 1 coil temperature, Sensor access port 1” diameter, Power supply breaker and convenience power switch located on facade, Door ajar alarm, audible and visual, Password protection – set-points and factory settings, 100 event alarm logging date/time stamp, Integrated Hi/Lo product temperature alarm test, Digital calibration air and product, Real time clock with date and time display, Adjustable alarm beep or constant on (key pad selectable), Alarm volume adjustable manually, Low battery test switch with buzzer.
Perimeter anti-condensate door frame (temperature selectable heater wire) , Sensor failure alarm, Power failure alarm, Exterior cabinet front, sides and back are powder coated, Exterior cabinet top and bottom are galvanized steel, Interior cabinet liner is stainless steel, Cabinet is foamed-in-place with CFC free high density, polyurethane foam insulation, Solid doors(s), full size, Heavy duty door pivot hinges, Pull door handles, full length, Magnetic door gaskets, Centered key door lock(s) (2 keys), Four casters on one and two door models (2 locking), Six casters (3 locking) on three door models, 3 coated wire shelves per door section, Interior drain with plug, Top mounted forced air refrigeration system, Condensate evaporation, LED lighting.
Exterior front, sides and back powder coated. Exterior top and bottom glavanized steel. Interior cabinet is stainless steel.
Certifications: UL, C-UL listed (60Hz models only).
Ordering information: Accessories not included. All Units include 18 months limited warranty on parts and labor (US & Canada). Warranties include 18 month parts and labor, 5 year compressor, (US and Canada), 18 months parts (International).