Filtron™ Surgical Mask, Off-The-Face/Anti-Fog Style, 3M™
Supplier: 3M Healthcare
1838R
56222-061BX 35.34 USD
56222-061
Filtron™ Surgical Mask, Off-The-Face/Anti-Fog Style, 3M™
Duckbill off the face construction helps reduce fogging and has vertical ties.
  • High performance surgical mask
Soft, breathable inner lining with level 2 fluid resistance which provides moderate fluid protection at 120 mmHg.