Would you like to log in again?
GeneMate Tissue Culture Flasks
Supplier: GeneMate
T-2000-2P T-2000-1P T-2000-5V T-2000-5P T-2000-4P T-2000-3P T-2000-4V T-2000-3V T-2000-2V T-2000-1V
  • Ideal for cell growth and cell yields
  • Angled neck offers sample access for pipetting and scraping
  • Flask surface is flat and free of striation to maximize usuable growth area
  • Volume graduations on two sides
  • Writing area near the neck for easy identification
  • Flasks come in a self-standing, resealable zip bag to ensure sterility
  • Sterilized by gamma irradiation; non-pyrogenic; integrity-tested
  • Made of crystal clear polystyrene
  • Stackable
Vented Caps feature a 0.22 µm hydrophobic filter enabling gas exchange and minimizing risk of contamination. Plug Seal Caps are suitable for use in both closed or open systems: a liquid/gas sheer seal exists when closed; unscrew the cap one-quarter turn to allow gas exchange.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Volume Surface Area VWR Catalog Number Unit Price Quantity
Flasks with Plug Seal Caps 250 mL 75 cm² 490017-094 Case of 100 Retrieving
Flasks with Plug Seal Caps 50 mL 25 cm² 490017-090 Case of 200 Retrieving
Flasks with Plug Seal Caps 600 mL 182.5 cm² 490017-098 Case of 40 Retrieving
Flasks with Plug Seal Caps 25 mL 12.5 cm² 490017-086 Case of 200 Retrieving
Flasks with Vented Caps 600 mL 182.5 cm² 490017-096 Case of 40 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty