Probenecid ≥98% (by titrimetric analysis)

Supplier: Enzo Life Sciences

Synonyms: 4-[(dipropylamino)sulphonyl]benzoic acid

430-113-G005
89151-130EA 52.99 USD
89151-130
Probenecid ≥98% (by titrimetric analysis)
Probenecid

Inhibitor of several ABC-transporters of the subfamily ABCC or MRP (multidrug resistant associated protein).

Formula: C₁₃H₁₉NO₄S
MW: 285.4 g/mol
MDL Number: MFCD00038402
CAS Number: 57-66-9
https://us.vwr.com/stibo/web/std.lang.all/06/81/45390681.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR