S-Nitrosocaptopril ≥98%
Supplier: Enzo Life Sciences
270-213-M050
89147-404EA
372.64
USD
89147-404
S-Nitrosocaptopril ≥98%
S-Nitrosocaptopril
Angiotensin-converting enzyme (ACE) inhibitor.
Inhibitor of platelet aggregation. Its activity may depend on the homolytic cleavage of the S-N bond under physiological conditions, yielding nitric oxide (NO) and the parent compound, captopril.
Formula:
C₉H₁₄N₂O₄S MW: 246.3 g/mol |
CAS Number:
122130-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...