3-(4-Hydroxybenzylidenyl)indolin-2-one ≥98%

270-363-M005
89147-708EA 313.16 USD
89147-708
3-(4-Hydroxybenzylidenyl)indolin-2-one ≥98%
3-(4-Hydroxybenzylidenyl)indolin-2-one

Selective receptor tyrosine kinase (RTK) inhibitor.


Inhibits FLK-1 RTK (IC50 ~3 µM). Consists predominantly of the E-isomer, contains ~10% of the Z-isomer.

Formula: C₁₅H₁₁NO₂
MW: 237.3 g/mol
CAS Number: 293302-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR