3-(3,5-Di-tert-butyl-4-hydroxybenzylidenyl)indolin-2-one ≥97% (by 1H-NMR, HPLC)

270-364-M005
89147-712EA 199.05 USD
89147-712
3-(3,5-Di-tert-butyl-4-hydroxybenzylidenyl)indolin-2-one ≥97% (by 1H-NMR, HPLC)
3-(3,5-Di-tert-butyl-4-hydroxybenzylidenyl)indolin-2-one

Receptor tyrosine kinase (RTK) inhibitor.


Inhibits FLK-1 RTK (IC50~8µM) and EGF RTK. Consists of the E- and Z-isomers.

Formula: C₂₃H₂₇NO₂
MW: 349.5 g/mol
CAS Number: 40526-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR