Samco™ Round Base Test Tubes, 16×125 mm, Thermo Scientific
Supplier: Thermo Scientific
16-002S 16-005
89498-466CS 1143.68 USD
89498-466 89498-460
Samco™ Round Base Test Tubes, 16×125 mm, Thermo Scientific
Tubes Test Tubes
Samco™ round based 16×125 mm tubes are suitable for most laboratory applications.
  • Available with or without cap
Available in clear polypropylene for chemical resistance and strength, or polystyrene for superior clarity.
Ordering information: Ambient storage and shipping recommended for tubes.
Packaging: Choice of packaging formats: sterile or non sterile.