Would you like to log in again?
Oxygen Scavengers Test, MilliporeSigma
Supplier: MilliporeSigma
Spectroquant®
1.19251.0001
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Range Chemical Method For Unit No. of Tests Supplier No. VWR Catalog Number Unit Price Quantity
0.020 - 0.500 mg/l DEHA 0.027 - 0.667 mg/l Carbohy 0.053 - 1.315 mg/l Hydro 0.078 - 1.950 mg/l ISA 0.087 - 2.170 mg/l MEKO Iron Reduction Prove, Nova 60, Multy 200 Tests 1.19251.0001 EM1.19251.0001 Each Retrieving Restricted
Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Storage Temperature Store at +15°C to +25°C.
Method Photometric 0.020 - 0.500 mg/l DEHA 0.027 - 0.667 mg/l Carbohy 0.053 - 1.315 mg/l Hydro 0.078 - 1.950 mg/l ISA 0.087 - 2.170 mg/l MEKO
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty