Programmable Electronic High-Power Crimp Stations, Thermo Scientific
ECRH-20KIT ECRH-11KIT ECRH-B
10144-760EA 1436.79 USD
10144-760 10144-762 10144-764
Programmable Electronic High-Power Crimp Stations, Thermo Scientific
Achieve secure, reproducible crimps with the push of a button by using programmable electronic high-power crimp stations
Decappers provide quick and easy removal of all types of seals with the push of a button. Adjustable crimp settings allow for compatibility with most vial/septum/seal combinations including aluminum, steel and bi-metal seals. Crimp-force sensing automatically determines when a proper seal has been formed and opens the jaws to release the vial. Higher power is perfect for steel caps. Exchangeable crimp and decrimp heads can be removed or installed in seconds. For each head, a set of up to 10 adjustment programs are available and can be saved. The fully electric operation eliminates the need for pressurized air supplies at the operation location.
A reliable crimp is guaranteed when programmed, with various closures, septa thickness and vials.
Ordering information: All models include the high-power crimper and 12V CD supply with power cord. Kits also include one each of the crimp and decrimp head of the listed size. Crimping/decapping jaw sets and an adjustable accessory base are available separately.