AlphaTec® 88-343 Natural Rubber Latex Gloves, Unlined, Natural, Ansell
Supplier: Ansell
AlphaTec®
114031
32890-637CS 417.33 USD
32890-637
AlphaTec® 88-343 Natural Rubber Latex Gloves, Unlined, Natural, Ansell
Gloves
An ideal glove for safety and dexterity in the food processing industry, while offering moderate chemical protection.
  • Great chemical protection
  • Excellent grip on wet or dry surfaces
  • Superior tensile strength and elasticity
  • Diamond embossed for excellent grip on wet or dry surfaces
The 20-millimeter thickness provides good chemical, snag, puncture, abrasion and cut resistance.
Use in the agriculture, viticulture, food processing, food service, and general industries. Uses include canning and food processing; food packing and handling; fruit and vegetable processing; light-duty maintenance; and clean up.
Certifications: Complies with FDA food handling requirements (21 CFR 177.2600).
Packaging: Gloves are packaged 12 pairs in a bag; 12 bags in a carton.